missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ D-Biotin

Catalog No. p-7075797
Encompass
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 mL
Catalog No. Quantity
B20656 10 mL
B1595 1 g
2 options

Catalog No. B20656

Supplier: Invitrogen™ B20656

Only null left

D-Biotin

D-biotin binds with high affinity to streptavidin, avidin and NeutrAvidin™ biotin-binding proteins. This solution can be used in conjunction with unlabeled streptavidin or avidin in procedures to block endogenous biotin.

Specifications

Concentration 50mM
Conjugate Unconjugated
Quantity 10 mL
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Room Temperature
Form Liquid
Product Line Invitrogen
Product Type Biotin (unconjugated)

For Research Use Only. Not for use in diagnostic procedures.