missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ D-Biotin

Catalog No. b20656
Encompass
Change view
Click to view available options
Quantity:
1 g
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20656 10 mL
B1595 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20656 Supplier Invitrogen™ Supplier No. B20656
Only null left

D-Biotin

D-biotin binds with high affinity to streptavidin, avidin and NeutrAvidin™ biotin-binding proteins. This solution can be used in conjunction with unlabeled streptavidin or avidin in procedures to block endogenous biotin.

Specifications

Concentration 50mM
Conjugate Unconjugated
Quantity 10 mL
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Room Temperature
Form Liquid
Product Line Invitrogen
Product Type Biotin (unconjugated)

For Research Use Only. Not for use in diagnostic procedures.