missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 1807350-90-8 |
|---|---|
| Molecular Formula | C7H14ClNO2 |
| Molecular Weight (g/mol) | 179.644 |
| MDL Number | MFCD18071037 |
| InChI Key | AYYKCALHTPKNOI-FYZOBXCZSA-N |
| Synonym | ethyl r-3-pyrrolidinecarboxylate hydrochloride, d-beta-proline ethyl ester hydrochloride, r-ethyl pyrrolidine-3-carboxylate hydrochloride, ethyl r-3-pyrrolidinecarboxylate, hcl, r-ethyl pyrrolidine-3-carboxylate hcl, ethyl r---pyrrolidine-3-carboxylate hydrochloride, ethyl r-pyrrolidine-3-carboxylate hydrochloride, ethyl 3r-pyrrolidine-3-carboxylate hydrochloride, r---pyrrolidine-3-carboxylic acid ethyl ester hydrochloride |
| PubChem CID | 53488453 |
| IUPAC Name | ethyl (3R)-pyrrolidine-3-carboxylate;hydrochloride |
| SMILES | CCOC(=O)C1CCNC1.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH5778903
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesProtect from heat. Store away from oxidizing agents. Refrigerate. Incompatible with heat and oxidizing agents.
Chemical Identifiers
| 1807350-90-8 | |
| 179.644 | |
| AYYKCALHTPKNOI-FYZOBXCZSA-N | |
| 53488453 | |
| CCOC(=O)C1CCNC1.Cl |
| C7H14ClNO2 | |
| MFCD18071037 | |
| ethyl r-3-pyrrolidinecarboxylate hydrochloride, d-beta-proline ethyl ester hydrochloride, r-ethyl pyrrolidine-3-carboxylate hydrochloride, ethyl r-3-pyrrolidinecarboxylate, hcl, r-ethyl pyrrolidine-3-carboxylate hcl, ethyl r---pyrrolidine-3-carboxylate hydrochloride, ethyl r-pyrrolidine-3-carboxylate hydrochloride, ethyl 3r-pyrrolidine-3-carboxylate hydrochloride, r---pyrrolidine-3-carboxylic acid ethyl ester hydrochloride | |
| ethyl (3R)-pyrrolidine-3-carboxylate;hydrochloride |
Specifications
| 1807350-90-8 | |
| MFCD18071037 | |
| AYYKCALHTPKNOI-FYZOBXCZSA-N | |
| ethyl (3R)-pyrrolidine-3-carboxylate;hydrochloride | |
| 53488453 | |
| −14° (c=0.6 in Water) | |
| D-beta-Proline ethyl ester hydrochloride |
| C7H14ClNO2 | |
| ethyl r-3-pyrrolidinecarboxylate hydrochloride, d-beta-proline ethyl ester hydrochloride, r-ethyl pyrrolidine-3-carboxylate hydrochloride, ethyl r-3-pyrrolidinecarboxylate, hcl, r-ethyl pyrrolidine-3-carboxylate hcl, ethyl r---pyrrolidine-3-carboxylate hydrochloride, ethyl r-pyrrolidine-3-carboxylate hydrochloride, ethyl 3r-pyrrolidine-3-carboxylate hydrochloride, r---pyrrolidine-3-carboxylic acid ethyl ester hydrochloride | |
| CCOC(=O)C1CCNC1.Cl | |
| 179.644 | |
| 179.65 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only