missing translation for 'onlineSavingsMsg'
Learn More

D-beta-Proline ethyl ester hydrochloride, 97%, Thermo Scientific™

Catalog No. AAH5778903
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5778903 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5778903 Supplier Thermo Scientific Chemicals Supplier No. H5778903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Protect from heat. Store away from oxidizing agents. Refrigerate. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 1807350-90-8
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD18071037
InChI Key AYYKCALHTPKNOI-FYZOBXCZSA-N
Synonym ethyl r-3-pyrrolidinecarboxylate hydrochloride, d-beta-proline ethyl ester hydrochloride, r-ethyl pyrrolidine-3-carboxylate hydrochloride, ethyl r-3-pyrrolidinecarboxylate, hcl, r-ethyl pyrrolidine-3-carboxylate hcl, ethyl r---pyrrolidine-3-carboxylate hydrochloride, ethyl r-pyrrolidine-3-carboxylate hydrochloride, ethyl 3r-pyrrolidine-3-carboxylate hydrochloride, r---pyrrolidine-3-carboxylic acid ethyl ester hydrochloride
PubChem CID 53488453
IUPAC Name ethyl (3R)-pyrrolidine-3-carboxylate;hydrochloride
SMILES CCOC(=O)C1CCNC1.Cl

Specifications

Quantity 1g
Formula Weight 179.65
Optical Rotation −14° (c=0.6 in Water)
Percent Purity 97%
Chemical Name or Material D-beta-Proline ethyl ester hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only