missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Aspartic acid, 99%

CAS: 1783-96-6 | C4H7NO4 | 133.103 g/mol

Supplier:  Thermo Scientific Chemicals B2118414

 View more versions of this product

Catalog No. AAB2118414


Only null left

Description

Description

D-Aspartic acid modulates melatonin synthesis in the pineal gland.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Aspartic acid modulates melatonin synthesis in the pineal gland.

Solubility
Slightly soluble in water, Soluble in 1N ammonium hydroxide (100 mg/ml), and dilute HCl. Insoluble in ether, and alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

>300°C (decomposition)
1783-96-6
MFCD00063081
14,840
Slightly soluble in water,Soluble in 1N ammonium hydroxide (100mg/ml),and dilute HCl. Insoluble in ether,and alcohol.
C(C(C(=O)O)N)C(=O)O
(2R)-2-aminobutanedioic acid
133.103
CHEBI:17364
133.1
D-Aspartic acid
C4H7NO4
1723529
d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d
CKLJMWTZIZZHCS-UWTATZPHSA-N
1.66
25 g
83887
−25° (c=1 in 1N HCl)
99%

Chemical Identifiers

1783-96-6
133.103
CKLJMWTZIZZHCS-UWTATZPHSA-N
83887
(2R)-2-aminobutanedioic acid
C4H7NO4
MFCD00063081
d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d
CHEBI:17364
C(C(C(=O)O)N)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-234-6

RTECSNumber : CI9097500

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only