missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Aspartic acid, 99%

Catalog No. p-7025605
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2118406 5 g
AAB2118414 25 g
AAB2118422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2118406 Supplier Thermo Scientific Chemicals Supplier No. B2118406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1783-96-6 | C4H7NO4 | 133.103 g/mol

D-Aspartic acid modulates melatonin synthesis in the pineal gland.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Aspartic acid modulates melatonin synthesis in the pineal gland.

Solubility
Slightly soluble in water, Soluble in 1N ammonium hydroxide (100 mg/ml), and dilute HCl. Insoluble in ether, and alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1783-96-6
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00063081
InChI Key CKLJMWTZIZZHCS-UWTATZPHSA-N
Synonym d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d
PubChem CID 83887
ChEBI CHEBI:17364
IUPAC Name (2R)-2-aminobutanedioic acid
SMILES C(C(C(=O)O)N)C(=O)O

Specifications

Melting Point >300°C (decomposition)
Beilstein 1723529
Merck Index 14,840
Solubility Information Slightly soluble in water,Soluble in 1N ammonium hydroxide (100mg/ml),and dilute HCl. Insoluble in ether,and alcohol.
Density 1.66
Quantity 5 g
Formula Weight 133.1
Optical Rotation −25° (c=1 in 1N HCl)
Percent Purity 99%
Chemical Name or Material D-Aspartic acid
EINECSNumber 217-234-6
RTECSNumber CI9097500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only