missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Aspartic acid, 99+%

CAS: 1783-96-6 | C4H7NO4 | 133.103 g/mol

Supplier:  Thermo Scientific Chemicals 204260250

 View more versions of this product

Catalog No. AC204260250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

>300.0°C
99+%
White
0.4% max. (105°C, 3 hrs)
0.3% max. (TLC)
Glass Bottle
MFCD00063081
− 25.80
15, 830
Solubility in water: soluble. Other solubilities: insoluble in alcohol and ether
C(C(C(=O)O)N)C(=O)O
133.103
83887
133.1
Crystalline Powder
D(-)-Aspartic acid
1783-96-6
Authentic
99+%
C4H7NO4
HO2CCH2CH(NH2)CO2H
0.1% max.
04, 471
d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d
CKLJMWTZIZZHCS-UWTATZPHSA-N
(2R)-2-aminobutanedioic acid
25 g
CHEBI:17364
≥99%

Chemical Identifiers

1783-96-6
133.103
CKLJMWTZIZZHCS-UWTATZPHSA-N
83887
(2R)-2-aminobutanedioic acid
C4H7NO4
MFCD00063081
d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d
CHEBI:17364
C(C(C(=O)O)N)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-234-6

RTECSNumber : CI9097500

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only