missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Aspartic acid, 99+%

Catalog No. p-61823
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC204260050 5 g Glass Bottle
AC204260250 25 g Glass Bottle
AC204261000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC204260050 Supplier Thermo Scientific Chemicals Supplier No. 204260050

CAS: 1783-96-6 | C4H7NO4 | 133.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1783-96-6
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00063081
InChI Key CKLJMWTZIZZHCS-UWTATZPHSA-N
Synonym d-aspartic acid, r-2-aminosuccinic acid, d-aspartate, h-d-asp-oh, r-aspartic acid, --aspartic acid, aspartic acid d-form, 2r-2-aminobutanedioic acid, d---aspartic acid, aspartic acid, d
PubChem CID 83887
ChEBI CHEBI:17364
IUPAC Name (2R)-2-aminobutanedioic acid
SMILES C(C(C(=O)O)N)C(=O)O

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.4% max. (105°C, 3 hrs)
Other Amino Acids 0.3% max. (TLC)
Packaging Glass Bottle
Linear Formula HO2CCH2CH(NH2)CO2H
Residue after Ignition 0.1% max.
Specific Rotation − 25.80
Beilstein 04, 471
Merck Index 15, 830
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol and ether
Specific Rotation Condition − 25.80 (20.00°C c=5,5N HCl)
Quantity 5 g
Formula Weight 133.1
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Aspartic acid, 99+%
Show More Show Less
EINECSNumber 217-234-6
RTECSNumber CI9097500

RUO – Research Use Only