missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Araboascorbic Acid 98.0+%, TCI America™
$38.51 - $90.48
Chemical Identifiers
| CAS | 89-65-6 |
|---|---|
| Molecular Formula | C6H7NaO6 |
| Molecular Weight (g/mol) | 198.11 |
| MDL Number | MFCD00005378 |
| InChI Key | IFVCRSPJFHGFCG-HXPAKLQESA-N |
| Synonym | erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5 |
| PubChem CID | 54675810 |
| ChEBI | CHEBI:51438 |
| IUPAC Name | sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide |
| SMILES | [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O |
Chemical Identifiers
| 89-65-6 | |
| 198.11 | |
| IFVCRSPJFHGFCG-HXPAKLQESA-N | |
| 54675810 | |
| sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide |
| C6H7NaO6 | |
| MFCD00005378 | |
| erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5 | |
| CHEBI:51438 | |
| [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O |
Specifications
| 89-65-6 | |
| White-Yellow | |
| MFCD00005378 | |
| erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5 | |
| [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O | |
| 198.11 | |
| CHEBI:51438 | |
| ≥98.0% (T) | |
| D-Araboascorbic Acid |
| 167°C | |
| C6H7NaO6 | |
| 25 g | |
| IFVCRSPJFHGFCG-HXPAKLQESA-N | |
| sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide | |
| 54675810 | |
| 176.12 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0131
RTECSNumber : KF3015000
TSCA : Yes