missing translation for 'onlineSavingsMsg'
Learn More

D-Araboascorbic Acid 98.0+%, TCI America™

Catalog No. A052025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A052025G 25 g
A0520500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A052025G Supplier TCI America Supplier No. A052025G

Chemical Identifiers

CAS 89-65-6
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00005378
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5
PubChem CID 54675810
ChEBI CHEBI:51438
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point 167°C
Color White-Yellow
Quantity 25 g
Formula Weight 176.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Araboascorbic Acid
EINECSNumber (9)-0131
RTECSNumber KF3015000
TSCA Yes