missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Arabitol 98.0+%, TCI America™

Catalog No. p-7116943
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A05161G 1 g
A051625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A05161G Supplier TCI America Supplier No. A05161G
Only null left

Chemical Identifiers

CAS 488-82-4
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.146
MDL Number MFCD00004709
InChI Key HEBKCHPVOIAQTA-QWWZWVQMSA-N
Synonym d-arabinitol, arabitol, dl-arabitol, d-arabitol, d-+-arabitol, d-+-arabinitol, d-lyxitol, lyxitol, dl-arabinitol, d-arabinol
PubChem CID 94154
ChEBI CHEBI:18333
IUPAC Name (2R,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

Melting Point 103°C
Color White
Quantity 1 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Arabitol