Learn More
Thermo Scientific Chemicals D-(+)-Arabitol, 99%
CAS: 488-82-4 | C5H12O5 | 152.146 g/mol
Supplier: Thermo Scientific Chemicals A1780114
Description
It can be employed as a sugar in the production of xylitol via biotransformation process. It is a substrate used to identify, differentiate and characterize enzyme such as the gluconobacter oxydans dehydrogenase(s), Gox2181, hyperthermophilic D-arabitol dehydrogenase from Thermotoga maritime and NAD-dependent D-arabitoldehydrogenase from acetic acid bacterium, Acetobacter suboxydans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be employed as a sugar in the production of xylitol via biotransformation process. It is a substrate used to identify, differentiate and characterize enzyme such as the gluconobacter oxydans dehydrogenase(s), Gox2181, hyperthermophilic D-arabitol dehydrogenase from Thermotoga maritime and NAD-dependent D-arabitoldehydrogenase from acetic acid bacterium, Acetobacter suboxydans.
Solubility
Soluble in water (50 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
Specifications
| D-(+)-Arabitol | |
| 100°C to 104°C | |
| MFCD00004709 | |
| 1720520 | |
| d-arabinitol, arabitol, dl-arabitol, d-arabitol, d-+-arabitol, d-+-arabinitol, d-lyxitol, lyxitol, dl-arabinitol, d-arabinol | |
| HEBKCHPVOIAQTA-QWWZWVQMSA-N | |
| C(C(C(C(CO)O)O)O)O | |
| 152.146 | |
| CHEBI:18333 | |
| 99% |
| 488-82-4 | |
| C5H12O5 | |
| 25 g | |
| 14,762 | |
| Soluble in water (50mg/ml). | |
| +132° (c=0.4 in Acidified Molybdate Solution) | |
| (2R,4R)-pentane-1,2,3,4,5-pentol | |
| 94154 | |
| 152.15 |
Chemical Identifiers
| 488-82-4 | |
| 152.146 | |
| HEBKCHPVOIAQTA-QWWZWVQMSA-N | |
| 94154 | |
| (2R,4R)-pentane-1,2,3,4,5-pentol |
| C5H12O5 | |
| MFCD00004709 | |
| d-arabinitol, arabitol, dl-arabitol, d-arabitol, d-+-arabitol, d-+-arabinitol, d-lyxitol, lyxitol, dl-arabinitol, d-arabinol | |
| CHEBI:18333 | |
| C(C(C(C(CO)O)O)O)O |
Safety and Handling
EINECSNumber : 207-686-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only