missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Arabitol, 99%

Catalog No. AAA1780106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1780106 5 g
AAA1780114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1780106 Supplier Thermo Scientific Chemicals Supplier No. A1780106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 488-82-4 | C5H12O5 | 152.146 g/mol

It can be employed as a sugar in the production of xylitol via biotransformation process. It is a substrate used to identify, differentiate and characterize enzyme such as the gluconobacter oxydans dehydrogenase(s), Gox2181, hyperthermophilic D-arabitol dehydrogenase from Thermotoga maritime and NAD-dependent D-arabitoldehydrogenase from acetic acid bacterium, Acetobacter suboxydans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be employed as a sugar in the production of xylitol via biotransformation process. It is a substrate used to identify, differentiate and characterize enzyme such as the gluconobacter oxydans dehydrogenase(s), Gox2181, hyperthermophilic D-arabitol dehydrogenase from Thermotoga maritime and NAD-dependent D-arabitoldehydrogenase from acetic acid bacterium, Acetobacter suboxydans.

Solubility
Soluble in water (50 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.

Chemical Identifiers

CAS 488-82-4
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.146
MDL Number MFCD00004709
InChI Key HEBKCHPVOIAQTA-QWWZWVQMSA-N
Synonym d-arabinitol, arabitol, dl-arabitol, d-arabitol, d-+-arabitol, d-+-arabinitol, d-lyxitol, lyxitol, dl-arabinitol, d-arabinol
PubChem CID 94154
ChEBI CHEBI:18333
IUPAC Name (2R,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

Melting Point 100°C to 104°C
Quantity 5 g
Beilstein 1720520
Merck Index 14,762
Solubility Information Soluble in water (50mg/ml).
Optical Rotation +132° (c=0.4 in Acidified Molybdate Solution)
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material D-(+)-Arabitol
EINECSNumber 207-686-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only