missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Arabinose, 99+%

Catalog No. p-60798
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161450250 25 g Plastic bottle
AC161451000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161450250 Supplier Thermo Scientific Chemicals Supplier No. 161450250
Only null left

CAS: 28697-53-2 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28697-53-2
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
InChI Key PYMYPHUHKUWMLA-DEOSMSJNSA-N
Synonym alpha-d-arabinopyranose, 2s,3s,4r,5r-oxane-2,3,4,5-tetrol, d--arabinose, alpha-arabinose, a-d-arabinopyranose, alpha-d-arabinopyranose 9ci, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
PubChem CID 71313478
IUPAC Name (4S,5R)-oxane-2,3,4,5-tetrol
SMILES OCC(O)[C@@H](O)[C@H](O)C=O

Specifications

Melting Point 152.0°C to 160.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 31, 34
Specific Rotation − 103.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glycerol and ether
Formula Weight 150.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material D(-)-Arabinose, p.a.
Show More Show Less

RUO – Research Use Only