missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-alpha-Tocopheryl acetate, 97%

Catalog No. AC421060010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC421060010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC421060010 Supplier Thermo Scientific Chemicals Supplier No. 421060010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58-95-7 | C31H52O3 | 472.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-95-7
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.75
MDL Number MFCD00072042,MFCD00072052
InChI Key ZAKOWWREFLAJOT-UHFFFAOYNA-N
Synonym vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
PubChem CID 86472
ChEBI CHEBI:32321
IUPAC Name 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydro-2H-1-benzopyran-6-yl acetate
SMILES CC(C)CCCC(C)CCCC(C)CCCC1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1

Specifications

Color Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4940 to 1.498
Quantity 1 g
Merck Index 15, 9653
Specific Rotation Condition +3° (25°C c=2,in ethanol)
Specific Rotation + 3
Formula Weight 472.75
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material D-α-Tocopheryl acetate, 98%
Show More Show Less
Hazard Category PRODUCT NEEDS INERT ATMOSPHERE
EINECSNumber 200-405-4

RUO – Research Use Only