missing translation for 'onlineSavingsMsg'
Learn More

D-alpha-Tocopherol Acetate 96.0+%, TCI America™

Catalog No. T232225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T232225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T232225G Supplier TCI America Supplier No. T232225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58-95-7
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.75
MDL Number MFCD00072042,MFCD00072052
InChI Key ZAKOWWREFLAJOT-UHFFFAOYNA-N
Synonym vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
PubChem CID 86472
ChEBI CHEBI:32321
IUPAC Name 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydro-2H-1-benzopyran-6-yl acetate
SMILES CC(C)CCCC(C)CCCC(C)CCCC1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 472.75
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material D-alpha-Tocopherol Acetate
EINECSNumber (9)-0487
RTECSNumber GP8280000
TSCA Yes