missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Allose 98.0+%, TCI America™
$124.89 - $659.87
Chemical Identifiers
| CAS | 2595-97-3 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | 180.156 |
| MDL Number | MFCD00135833 |
| InChI Key | GZCGUPFRVQAUEE-OBOOZECYSA-N |
| PubChem CID | 57449163 |
| IUPAC Name | (2R,3R,4R)-2,3,4,5,6-pentahydroxyhexanal |
| SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1488100MG
|
TCI America
A1488100MG |
100 mg |
Each for $124.89
|
|
|||||
|
A14881G
|
TCI America
A14881G |
1 g |
Each for $659.87
|
|
|||||
Chemical Identifiers
| 2595-97-3 | |
| 180.156 | |
| GZCGUPFRVQAUEE-OBOOZECYSA-N | |
| (2R,3R,4R)-2,3,4,5,6-pentahydroxyhexanal |
| C6H12O6 | |
| MFCD00135833 | |
| 57449163 | |
| C(C(C(C(C(C=O)O)O)O)O)O |
Specifications
| 2595-97-3 | |
| White | |
| MFCD00135833 | |
| GZCGUPFRVQAUEE-OBOOZECYSA-N | |
| (2R,3R,4R)-2,3,4,5,6-pentahydroxyhexanal | |
| 57449163 | |
| ≥98.0% (HPLC) | |
| D-(+)-Allose |
| 149°C | |
| C6H12O6 | |
| 100 mg | |
| C(C(C(C(C(C=O)O)O)O)O)O | |
| 180.156 | |
| 180.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No