missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Allose 98.0+%, TCI America™

Catalog No. A1488100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1488100MG 100 mg
A14881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1488100MG Supplier TCI America Supplier No. A1488100MG

Chemical Identifiers

CAS 2595-97-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00135833
InChI Key GZCGUPFRVQAUEE-OBOOZECYSA-N
PubChem CID 57449163
IUPAC Name (2R,3R,4R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 149°C
Color White
Quantity 100 mg
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Allose