missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Alanine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
$46.60 - $128.62
Chemical Identifiers
| CAS | 59531-86-1 |
|---|---|
| Molecular Formula | C7H16ClNO2 |
| Molecular Weight (g/mol) | 181.66 |
| MDL Number | MFCD00077087 |
| InChI Key | WIQIWPPQGWGVHD-NUBCRITNSA-N |
| Synonym | d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride |
| PubChem CID | 16211919 |
| IUPAC Name | tert-butyl (2R)-2-aminopropanoate;hydrochloride |
| SMILES | CC(C(=O)OC(C)(C)C)N.Cl |
Chemical Identifiers
| 59531-86-1 | |
| 181.66 | |
| WIQIWPPQGWGVHD-NUBCRITNSA-N | |
| 16211919 | |
| CC(C(=O)OC(C)(C)C)N.Cl |
| C7H16ClNO2 | |
| MFCD00077087 | |
| d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride | |
| tert-butyl (2R)-2-aminopropanoate;hydrochloride |
Specifications
| 59531-86-1 | |
| White-Yellow | |
| MFCD00077087 | |
| d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride | |
| CC(C(=O)OC(C)(C)C)N.Cl | |
| 181.66 | |
| 181.66 | |
| Crystalline Powder |
| 208°C | |
| C7H16ClNO2 | |
| 1 g | |
| WIQIWPPQGWGVHD-NUBCRITNSA-N | |
| tert-butyl (2R)-2-aminopropanoate;hydrochloride | |
| 16211919 | |
| ≥98.0% (HPLC,N) | |
| D-Alanine tert-Butyl Ester Hydrochloride |
Safety and Handling
TSCA : No