accessibility menu, dialog, popup

UserName

CAS RN 59531-86-1

CH3OOCH3H3CCH3H2NHCl

CAS RN 59531-86-1

IUPAC Name: tert-butyl 2-aminopropanoate hydrochloride
Synonyms: tert-butyl 2-aminopropanoate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: WIQIWPPQGWGVHD-UHFFFAOYNA-N
SMILES: Cl.CC(N)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)