missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine 98.0+%, TCI America™

Supplier:  TCI America A01775G

 View more versions of this product

Catalog No. A01775G


Only null left

Description

Description

Specifications

Specifications

D-Alanine
White
MFCD00008077
QNAYBMKLOCPYGJ-UWTATZPHSA-N
(2R)-2-aminopropanoic acid
89.09
CHEBI:15570
≥98.0% (T)
338-69-2
C3H7NO2
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
C[C@@H](N)C(O)=O
5 g
71080
89.09
Crystalline Powder

Chemical Identifiers

338-69-2
89.09
QNAYBMKLOCPYGJ-UWTATZPHSA-N
71080
(2R)-2-aminopropanoic acid
C3H7NO2
MFCD00008077
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
CHEBI:15570
C[C@@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1553

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions