missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine 98.0+%, TCI America™

Catalog No. p-7116761
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01775G 5 g
A017725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A01775G Supplier TCI America Supplier No. A01775G
Only null left

Chemical Identifiers

CAS 338-69-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008077
InChI Key QNAYBMKLOCPYGJ-UWTATZPHSA-N
Synonym d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
PubChem CID 71080
ChEBI CHEBI:15570
IUPAC Name (2R)-2-aminopropanoic acid
SMILES C[C@@H](N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 89.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Alanine
EINECSNumber (9)-1553
TSCA Yes