missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Alanine 98.0+%, TCI America™
Supplier: TCI America A017725G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C3H7NO2 | |
| MFCD00008077 | |
| d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form | |
| CHEBI:15570 | |
| C[C@@H](N)C(O)=O |
Specifications
| D-Alanine | |
| White | |
| MFCD00008077 | |
| QNAYBMKLOCPYGJ-UWTATZPHSA-N | |
| (2R)-2-aminopropanoic acid | |
| 25 g | |
| CHEBI:15570 | |
| ≥98.0% (T) |
| 338-69-2 | |
| C3H7NO2 | |
| d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form | |
| C[C@@H](N)C(O)=O | |
| 89.09 | |
| 71080 | |
| 89.09 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1553
TSCA : Yes