missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Alanine ethyl ester hydrochloride, 95%
CAS: 6331-09-5 | C8H19NO2 | 161.25 g/mol
$78.92 - $298.34
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6279106
|
Thermo Scientific Chemicals
H6279106 |
5 g |
Each for $78.92
|
|
|||||
|
AAH6279114
|
Thermo Scientific Chemicals
H6279114 |
25 g |
Each for $298.34
|
|
|||||
Description
D-Alanine ethyl ester hydrochloride is used as organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Alanine ethyl ester hydrochloride is used as organic intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| 6331-09-5 | |
| MFCD00190725 |
Specifications
| 6331-09-5 | |
| MFCD00190725 | |
| CYHRNDXVTAWKMO-UHFFFAOYNA-N | |
| ethyl (2R)-2-aminopropanoate;hydrochloride | |
| 152.60 | |
| 153.61 | |
| D-Alanine ethyl ester hydrochloride |
| C5H11ClNO2 | |
| d-alanine ethyl ester hydrochloride, r-ethyl 2-aminopropanoate hydrochloride, h-d-ala-oet hcl, r-ethyl 2-amino-propionate hcl, ethyl 2r-2-aminopropanoate hydrochloride, h-ala-oblz. tos, h-d-ala-oet. hcl, 3-alanine ethyl ester hydrochloride, r-ethyl 2-amino-propionate hydrochloride | |
| CCOC(=O)C(C)N.Cl | |
| 5 g | |
| 10236019 | |
| 95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only