Learn More
D-Alanine ethyl ester hydrochloride, 95%
CAS: 6331-09-5 | C8H19NO2 | 161.25 g/mol
Supplier: Thermo Scientific Chemicals H6279114
Description
D-Alanine ethyl ester hydrochloride is used as organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Alanine ethyl ester hydrochloride is used as organic intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
| D-Alanine ethyl ester hydrochloride | |
| C8H19NO2 | |
| 4,4-diethoxybutylamine, 4-aminobutyraldehyde diethyl acetal, 1-butanamine, 4,4-diethoxy, 4,4-diethoxy-1-butanamine, ambda, 4,4-diethoxy-butylamine, acmc-2097jf, 4-aminobutanal diethyl acetal, ksc356g0p, 4,4-diethoxy-1-butanamine # | |
| CCOC(CCCN)OCC | |
| 25 g | |
| 80672 | |
| 95% |
| 6331-09-5 | |
| MFCD00190725 | |
| GFLPSABXBDCMCN-UHFFFAOYSA-N | |
| 4,4-diethoxybutan-1-amine | |
| 161.25 | |
| 153.61 |
Chemical Identifiers
| 6331-09-5 | |
| MFCD00190725 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only