missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-4-Hydroxyphenylglycine, 98+%

Catalog No. ac201320250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC201320250 25 g Plastic bottle
AC201321000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC201320250 Supplier Thermo Scientific Chemicals Supplier No. 201320250
Only null left

CAS: 22818-40-2 | C8H9NO3 | 167.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22818-40-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00004262
InChI Key LJCWONGJFPCTTL-SSDOTTSWSA-N
Synonym d-4-hydroxyphenylglycine, 4-hydroxy-d-phenylglycine, d--4-hydroxyphenylglycine, h-d-phg 4-oh-oh, d-n-4-hydroxyphenyl glycine, 2r-2-amino-2-4-hydroxyphenyl acetic acid, r-2-amino-2-4-hydroxyphenyl acetic acid, r-alpha-amino-4-hydroxybenzeneacetic acid, d---4-hydroxyphenylglycine, unii-pcm9oix717
PubChem CID 89853
ChEBI CHEBI:15695
IUPAC Name (2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)O

Specifications

Melting Point 240°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula HOC6H4CH(NH2)CO2H
Quantity 25 g
Beilstein 14, I, 659
Specific Rotation Condition −156° (20°C c=1,1 N HCl)
Specific Rotation -156
Solubility Information Solubility in water: 5g/L (20 c)
Formula Weight 167.16
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material D(-)-4-Hydroxyphenylglycine, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 245-247-7
TSCA TSCA

RUO – Research Use Only