missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-2-Aminoadipic Acid 95.0+%, TCI America™
$95.06 - $210.63
Chemical Identifiers
| CAS | 7620-28-2 |
|---|---|
| Molecular Formula | C6H11NO4 |
| Molecular Weight (g/mol) | 161.157 |
| MDL Number | MFCD00063118 |
| InChI Key | OYIFNHCXNCRBQI-SCSAIBSYSA-N |
| Synonym | d-2-aminoadipic acid, d-homoglutamic acid, r-2-aminohexanedioic acid, d-a-aminoadipic acid, d-, d-, r-2-aminoadipic acid, r-2-amino-hexanedioic acid, r-2-amino-hexanedioic acid, hexanedioic acid, 2-amino-, 2r, hexanedioic acid, 2-amino-, 2r |
| PubChem CID | 165627 |
| ChEBI | CHEBI:37025 |
| IUPAC Name | (2R)-2-aminohexanedioic acid |
| SMILES | C(CC(C(=O)O)N)CC(=O)O |
Chemical Identifiers
| 7620-28-2 | |
| 161.157 | |
| OYIFNHCXNCRBQI-SCSAIBSYSA-N | |
| 165627 | |
| (2R)-2-aminohexanedioic acid |
| C6H11NO4 | |
| MFCD00063118 | |
| d-2-aminoadipic acid, d-homoglutamic acid, r-2-aminohexanedioic acid, d-a-aminoadipic acid, d-, d-, r-2-aminoadipic acid, r-2-amino-hexanedioic acid, r-2-amino-hexanedioic acid, hexanedioic acid, 2-amino-, 2r, hexanedioic acid, 2-amino-, 2r | |
| CHEBI:37025 | |
| C(CC(C(=O)O)N)CC(=O)O |
Specifications
| 7620-28-2 | |
| White-Yellow | |
| MFCD00063118 | |
| d-2-aminoadipic acid, d-homoglutamic acid, 2r-2-aminohexanedioic acid, 2r-2-aminohexanedioic acid, d-, d-, a-aminoadipic acid, r-2-aminoadipic acid, hexanedioic acid, 2-amino-,--and +, hexanedioic acid, 2-amino-, 2r, hexanedioic acid, 2-amino-, 2r | |
| C(CC(C(=O)O)N)CC(=O)O | |
| 161.157 | |
| CHEBI:37025 | |
| ≥95.0% (GC,T) | |
| D-2-Aminoadipic Acid |
| 187°C | |
| C6H11NO4 | |
| 1 g | |
| OYIFNHCXNCRBQI-SCSAIBSYSA-N | |
| (2R)-2-aminohexanedioic acid | |
| 165627 | |
| 161.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No