missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytomegalovirus gH Late Antigen Recombinant Human Monoclonal Antibody (MSL-109)
GREENER_CHOICE

Catalog No. MA551801
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551801 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551801 Supplier Invitrogen™ Supplier No. MA551801
Only null left

Human Recombinant Monoclonal Antibody

Binds human cytomegalovirus (CMV).

Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Cytomegalovirus glycoprotein H (gH) late neutralizing antigen and reacts with MRC-5 infected cells.

Specifications

Antigen Cytomegalovirus gH Late Antigen
Applications Neutralization
Classification Recombinant Monoclonal
Clone MSL-109
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias CMV
Host Species Human
Immunogen CMV antigen (Virus antigen batch V2547) prepared from MRC-5 cells infected with the Towne strain of CMV.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less