missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytomegalovirus gH Late Antigen Chimeric Recombinant Rabbit Monoclonal Antibody (HCMV16)
GREENER_CHOICE

Catalog No. MA551608
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551608 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551608 Supplier Invitrogen™ Supplier No. MA551608
Only null left

Rabbit Recombinant Monoclonal Antibody

This antibody binds gpUL75 (gH) of the human cytomegalovirus, which is an 86-kDa glycoprotein. The heterodimer glycoprotein H-glycoprotein L is required for the fusion of viral and plasma membranes leading to virus entry into the host cell. It is though to bind a conformational epitope on the gH, and its bind is destroyed if the protein is denatured.

Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Cytomegalovirus glycoprotein H (gH) late neutralizing antigen and reacts with MRC-5 infected cells.

Specifications

Antigen Cytomegalovirus gH Late Antigen Chimeric
Applications ELISA, Immunoprecipitation, Neutralization, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HCMV16
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias CMV
Host Species Rabbit
Immunogen The original antibody was generated by immunization of mice with purified HCMV strain AD169.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less