Learn More
StatLab Cytology Double Funnel Kit
Disposable funnel offers a lower risk of contamination than reusable funnels.
Supplier: StatLab STLM96410FW
Description
- Pre-attached filter card for reliable and consistent results
- For sample volumes up to 0.5 mL
- Can be used with all stainless steel slide clips
- 500 per case
Specifications
| 0.5 mL | |
| Clear | |
| Cytology Funnel |
| Polypropylene | |
| Single Funnel w/ Filter Card and Cap |