Learn More
StatLab Cytology Double Funnel Kit
Disposable funnel offers a lower risk of contamination than reusable funnels.
$2,884.82 - $3,639.75
Specifications
| Capacity (Metric) | 0.5 mL |
|---|---|
| Material | Polypropylene |
| Color | Clear |
| Product Type | Cytology Funnel |
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
|
STLM96420FW
|
StatLab
STLM96420FW |
Double Funnel w/ Filter Card and Cap |
Case of 500 Each for $3,504.93
|
|
|||||
|
STLM96510FW
|
StatLab
STLM96510FW |
Single Funnel w/ White Filter Card, Cap and Clip |
Case of 480 Each for $3,639.75
|
|
|||||
|
STLM96410FW
|
StatLab
STLM96410FW |
Single Funnel w/ Filter Card and Cap |
Case of 500 Each for $2,884.82
|
|
|||||
Description
- Pre-attached filter card for reliable and consistent results
- For sample volumes up to 0.5 mL
- Can be used with all stainless steel slide clips
- 500 per case
Specifications
| 0.5 mL | |
| Polypropylene | |
| Clear | |
| Cytology Funnel |