Learn More
StatLab Cytology Double Funnel Kit
Catalog No. STLM96420FW
Change view
Click to view available options
Includes:
Double Funnel w/ Filter Card and Cap
Single Funnel w/ Filter Card and Cap
Single Funnel w/ White Filter Card, Cap and Clip
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
STLM96420FW Double Funnel w/ Filter Card and Cap
STLM96510FW Single Funnel w/ White Filter Card, Cap and Clip
STLM96410FW Single Funnel w/ Filter Card and Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. STLM96420FW Supplier StatLab Supplier No. STLM96420FW
Only null left
Disposable funnel offers a lower risk of contamination than reusable funnels.
  • Pre-attached filter card for reliable and consistent results
  • For sample volumes up to 0.5 mL
  • Can be used with all stainless steel slide clips
  • 500 per case
Specifications
Capacity (Metric) 0.5 mL
Material Polypropylene
Color Clear
Includes Double Funnel w/ Filter Card and Cap
Product Type Cytology Funnel