missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytokeratin 4 Monoclonal Antibody (K06324_3A1), Invitrogen™

Catalog No. p-200088940
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555950 50 μL
MA556330 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555950 Supplier Invitrogen™ Supplier No. MA555950
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: ISTTTLNKRR

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between approximately40 kDa and approximately68 kDa and isoelectric pH between 4. 9-7. 8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.

Specifications

Antigen Cytokeratin 4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K06324_3A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene KRT4
Gene Accession No. P19013
Gene Alias 57kd keratin; 57kDa keratin; AW108092; cb59; CK4; CK-4; CYK4; cytokeratin; cytokeratin 4; cytokeratin-4; Cytoskeletal 57 kDa keratin; etID20321.6; intermediate filament protein; K4; Kb4; keratin 4; keratin 4, type II; keratin 8; keratin complex 2, basic, gene 4; keratin, type II cytoskeletal 4; Keratin-4; Krt-2.4; Krt2-4; Krt4; krt4 {ECO:0000250; krt4 protein; krt8; type II basic cytokeratin; type II keratin Kb4; type-II keratin Kb4; UniProtKB:P19013}; WSN1; wu:fa91a11; wu:fd19e09; wu:fl23c09; zf-K8
Gene Symbols KRT4
Host Species Mouse
Immunogen Human KRT4 peptide (immunogen range is 511-520).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3851
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less