missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytokeratin 23 Monoclonal Antibody (K56004_3E9), Invitrogen™

Catalog No. MA555746
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555746 50 μL
MA556126 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555746 Supplier Invitrogen™ Supplier No. MA555746
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: GTREESKSSM KVSATPKIKA

The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.

Specifications

Antigen Cytokeratin 23
Applications Western Blot
Classification Monoclonal
Clone K56004_3E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene KRT23
Gene Accession No. Q9C075
Gene Alias C HAIK1; CK23; CK-23; cytokeratin 23; cytokeratin-23; DKFZP434G032; HAIK1; histone deacetylase inducible keratin 23; hyperacetylation-inducible type I keratin; K23; Ka23; keratin 23; keratin 23 (histone deacetylase inducible); keratin 23, type I; keratin complex 1, acidic, gene 23; keratin, type I cytoskeletal 23; Keratin-23; Krt1-23; KRT23; MGC26158; type I intermediate filament cytokeratin; type I keratin KA23
Gene Symbols KRT23
Host Species Mouse
Immunogen Human KRT23 peptide (immunogen range is 381-400).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less