Learn More
Invitrogen™ Cytokeratin 18 Monoclonal Antibody (C-04), FITC
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA110326
Catalog No. PIMA110326
Only null left
Description
Description
This antibody reacts with cytokeratin 18 (45 kDa intracellular antigen), a member of intermediate filaments subfamily represented in epithelial tissues.
Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
Specifications
Chemical Identifiers
Cytokeratin 18
0.1 mg/mL
Flow Cytometry, Immunocytochemistry
FITC
Mouse
RUO
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P05783, P05784, Q5BJY9
16668, 294853, 3875
Cytoskeleton preparation of epidermal carcinoma cell line A431.
Antibody
C-04
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG1
Human, Mammalia, Mouse, Rat
KRT18
cb83; Cell proliferation-inducing gene 46 protein; CK 18; CK18; CK-18; CYK18; cytokeratin 18; cytokeratin endo B; cytokeratin type -1; DAPK-1; Cytokeratin18; cytokeratin-18; dapk1; dorsal aorta proneprin kinesin-1; DreK18; Endo B; I79_023185; K 18; K18; keratin 18; keratin 18, type I; keratin 18a, tandem duplicate 1; keratin complex 1, acidic, gene 18; keratin D; Keratin, type I cytoskeletal 18; Keratin18; keratin-18; Kerd; KRT 18; Krt1-18; KRT18; krt18 {ECO:0000250; krt18a.1; LOC100008885; PIG46; sb:cb83; type I cytoskeletal 18; UniProtKB:P05783}; wu:fa13f02; wu:fb36b04
KRT18
Primary
Size-exclusion chromatography
Specifications
Cytokeratin 18
Monoclonal
0.1 mg/mL
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P05783, P05784, Q5BJY9
KRT18
Cytoskeleton preparation of epidermal carcinoma cell line A431.
100 μg
Primary
Human, Mammalia, Mouse, Rat
Antibody
IgG1
Flow Cytometry, Immunocytochemistry
C-04
FITC
KRT18
cb83; Cell proliferation-inducing gene 46 protein; CK 18; CK18; CK-18; CYK18; cytokeratin 18; cytokeratin endo B; cytokeratin type -1; DAPK-1; Cytokeratin18; cytokeratin-18; dapk1; dorsal aorta proneprin kinesin-1; DreK18; Endo B; I79_023185; K 18; K18; keratin 18; keratin 18, type I; keratin 18a, tandem duplicate 1; keratin complex 1, acidic, gene 18; keratin D; Keratin, type I cytoskeletal 18; Keratin18; keratin-18; Kerd; KRT 18; Krt1-18; KRT18; krt18 {ECO:0000250; krt18a.1; LOC100008885; PIG46; sb:cb83; type I cytoskeletal 18; UniProtKB:P05783}; wu:fa13f02; wu:fb36b04
Mouse
Size-exclusion chromatography
RUO
16668, 294853, 3875
4°C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions
Product Suggestions