missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytochrome P450 Reductase Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA129792
Description
Suggested positive controls for this product are rat liver, NADPH, cytochrome, P450 and reductase protein.
This gene encodes an endoplasmic reticulum membrane oxidoreductase with an FAD-binding domain and a flavodoxin-like domain. The protein binds two cofactors, FAD and FMN, which allow it to donate electrons directly from NADPH to all microsomal P450 enzymes. Mutations in this gene have been associated with various diseases, including apparent combined P450C17 and P450C21 deficiency, amenorrhea and disordered steroidogenesis, congenital adrenal hyperplasia and Antley-Bixler syndrome.Chemical Identifiers
| Cytochrome P450 Reductase | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Sheep, Hamster, Pig, Guinea Pig | |
| POR | |
| 4933424M13Rik; CCR; CPR; CY DKFZp686G04235; CYPOR; cytochrome p450 oxidoreductase; cytochrome P450 reductase; DKFZp686G04235; FLJ26468; I79_007261; LOC101115252; NADPH Cytochrome; NADPH-cytochrome P450 oxidoreductase; NADPH-cytochrome P-450 oxidoreductase; NADPH-cytochrome P450 reductase; NADPH--cytochrome P450 reductase; NADPH-cytochrome P-450 reductase; NADPH-dependent cytochrome P450 reductase; P450 (cytochrome) oxidoreductase; P450 Reductase; P450R; Por; unnamed protein product | |
| POR | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.09% sodium azide | |
| P00388, P00389, P04175, P16435, P37039, P37040 | |
| 100135602, 100170114, 100301554, 100689241, 101115252, 18984, 29441, 489816, 5447 | |
| Purified rat liver NADPH-Cytochrome P450 reductase. | |
| Antibody |
Specifications
| Cytochrome P450 Reductase | |
| Polyclonal | |
| Unconjugated | |
| POR | |
| 4933424M13Rik; CCR; CPR; CY DKFZp686G04235; CYPOR; cytochrome p450 oxidoreductase; cytochrome P450 reductase; DKFZp686G04235; FLJ26468; I79_007261; LOC101115252; NADPH Cytochrome; NADPH-cytochrome P450 oxidoreductase; NADPH-cytochrome P-450 oxidoreductase; NADPH-cytochrome P450 reductase; NADPH--cytochrome P450 reductase; NADPH-cytochrome P-450 reductase; NADPH-dependent cytochrome P450 reductase; P450 (cytochrome) oxidoreductase; P450 Reductase; P450R; Por; unnamed protein product | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100135602, 100170114, 100301554, 100689241, 101115252, 18984, 29441, 489816, 5447 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide | |
| P00388, P00389, P04175, P16435, P37039, P37040 | |
| POR | |
| Purified rat liver NADPH-Cytochrome P450 reductase. | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Sheep, Hamster, Pig, Guinea Pig | |
| Antibody | |
| IgG |