missing translation for 'onlineSavingsMsg'
Learn More
Learn More
0.8 Percent Platinum on Carbon Powder, Reduced, Water Wet, 25 microns, BASF Catalyst, Spectrum™ Chemical
Specifications
Specifications
| Physical Form | Powder or Liquid |
| Quantity | 100 g |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | 0.8% Platinum on Carbon Powder, Reduced, Water Wet, 25 μ, BASF Catalyst |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.