missing translation for 'onlineSavingsMsg'
Learn More

Cytochalasin B

An actin-disrupting agent | CAS: 14930-96-2 | C29H37NO5 | 479.62 g/mol

Supplier:  Thermo Scientific Chemicals J65342LB0

 View more versions of this product

Catalog No. AAJ65342LB0


Only null left
Explore available promotions

Description

Description

Cytochalasin B is used for characterizing some of the polymerization properties of actin, and in studies on cytokinesis. Probe for the two hexose-transport systems in rat L6 myoblasts. And induces DNA fragmentation, inhibits cell division, and disrupts many cell processes. Inhibits glucose transport.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytochalasin B is used for characterizing some of the polymerization properties of actin, and in studies on cytokinesis. Probe for the two hexose-transport systems in rat L6 myoblasts. And induces DNA fragmentation, inhibits cell division, and disrupts many cell processes. Inhibits glucose transport.

Solubility
Freely soluble in DMF, DMSO or ethanol at room temperature

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Cytochalasin B
218°C to 221°C
Odorless
MFCD00077704
UN1544
14,2790
Freely soluble in DMF, DMSO or ethanol at room temperature
CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C
479.62
479.61
14930-96-2
White
C29H37NO5
5 mg
1096207
cytochalasin b, cytochalasin b from helminthosporium dematioideum
GBOGMAARMMDZGR-UHFFFAOYNA-N
16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione
44634701
Powder

Chemical Identifiers

14930-96-2
479.62
GBOGMAARMMDZGR-UHFFFAOYNA-N
44634701
CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C
C29H37NO5
MFCD00077704
cytochalasin b, cytochalasin b from helminthosporium dematioideum
16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H361
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Suspected of damaging fertility or the unborn child.

P201-P202-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P302+P350-P304+P340-P308+P313-P310-P330-P361-P363-P501c

H300+H310+H330-H361

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 239-000-2

RTECSNumber : RO0205000

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only