Learn More
Thermo Scientific Chemicals Cytochalasin B
Cytochalasin B, CAS# 14930-96-2, also known as phomin, is an organic heterotricyclic compound that is a cell-permeable mycotoxin with potent activity to inhibit cytoplasmic division. | CAS: 14930-96-2 | C29H37NO5 | 479.62 g/mol
$141.15 - $2270.99
Chemical Identifiers
| CAS | 14930-96-2 |
|---|---|
| Molecular Formula | C29H37NO5 |
| Molecular Weight (g/mol) | 479.62 |
| MDL Number | MFCD00077704 |
| InChI Key | GBOGMAARMMDZGR-UHFFFAOYNA-N |
| Synonym | cytochalasin b, cytochalasin b from helminthosporium dematioideum |
| PubChem CID | 44634701 |
| IUPAC Name | 16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione |
| SMILES | CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC228090010
|
Thermo Scientific Chemicals
228090010 |
1 mg | Glass bottle |
Each for $141.15
|
|
||||
|
AC228090050
|
Thermo Scientific Chemicals
228090050 |
5 mg | Glass bottle |
Each for $568.38
|
|
||||
|
AC228090250
|
Thermo Scientific Chemicals
228090250 |
25 mg | Glass bottle |
Each for $2,270.99
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• Cytochalasin B is a cell-permeable fungal toxin / mycotoxin considered to be an inhibitor of cytoplasmic division by blocking the formation of contractile microfilaments
Applications
• Cytochalasin B can inhibit platelet aggregation and actin polymerization
• Cytochalasin B is commonly used in studies on cytokinesis due to its ability to act as an inhibitor
• It is used for characterizing some of the polymerization properties of actin
• This compound can also induce DNA fragmentation, inhibit cell division, and inhibit glucose transport, among other molecular processes
• It could be used as a probe for the two hexose-transport systems in rat L6 myoblasts
• Cytochalasin B is commonly used in studies of glucose transporters (GLUT)
• It can be used in in vitro micronucleus assay protocols
Chemical Identifiers
| 14930-96-2 | |
| 479.62 | |
| GBOGMAARMMDZGR-UHFFFAOYNA-N | |
| 44634701 | |
| CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C |
| C29H37NO5 | |
| MFCD00077704 | |
| cytochalasin b, cytochalasin b from helminthosporium dematioideum | |
| 16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione |
Specifications
| 14930-96-2 | |
| White | |
| Glass bottle | |
| MFCD00077704 | |
| 15, 2787 | |
| cytochalasin b, cytochalasin b from helminthosporium dematioideum | |
| GBOGMAARMMDZGR-UHFFFAOYNA-N | |
| 16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione | |
| 44634701 | |
| ≥98% | |
| Cytochalasin B |
| 218.0°C to 223.0°C | |
| Authentic | |
| C29H37NO5 | |
| 1 mg | |
| + 89.00 | |
| Solubility in water: insoluble. Other solubilities: 10.3 mg/mL acetone - 35.4 mg/mL ethanol (24 c), 371 mg/mL dmso - 492 mg/mL dmf (24 c) | |
| CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C | |
| 479.62 | |
| 479.6 | |
| Powder |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Suspected of damaging fertility or the unborn child if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
EINECSNumber : 239-000-2
RUO – Research Use Only