missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytochalasin B

Catalog No. AC228090250
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228090250 25 mg Glass bottle
AC228090010 1 mg Glass bottle
AC228090050 5 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC228090250 Supplier Thermo Scientific Chemicals Supplier No. 228090250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cytochalasin B, CAS# 14930-96-2, also known as phomin, is an organic heterotricyclic compound that is a cell-permeable mycotoxin with potent activity to inhibit cytoplasmic division. | CAS: 14930-96-2 | C29H37NO5 | 479.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Cytochalasin B is a cell-permeable fungal toxin / mycotoxin considered to be an inhibitor of cytoplasmic division by blocking the formation of contractile microfilaments

Applications

• Cytochalasin B can inhibit platelet aggregation and actin polymerization
• Cytochalasin B is commonly used in studies on cytokinesis due to its ability to act as an inhibitor
• It is used for characterizing some of the polymerization properties of actin
• This compound can also induce DNA fragmentation, inhibit cell division, and inhibit glucose transport, among other molecular processes
• It could be used as a probe for the two hexose-transport systems in rat L6 myoblasts
• Cytochalasin B is commonly used in studies of glucose transporters (GLUT)
• It can be used in in vitro micronucleus assay protocols

Chemical Identifiers

CAS 14930-96-2
Molecular Formula C29H37NO5
Molecular Weight (g/mol) 479.62
MDL Number MFCD00077704
InChI Key GBOGMAARMMDZGR-UHFFFAOYNA-N
Synonym cytochalasin b, cytochalasin b from helminthosporium dematioideum
PubChem CID 44634701
IUPAC Name 16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione
SMILES CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C

Specifications

Melting Point 218.0°C to 223.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 mg
Merck Index 15, 2787
Specific Rotation Condition +89.00 (20.00°C c=1,CH3OH)
Specific Rotation + 89.00
Solubility Information Solubility in water: insoluble. Other solubilities: 10.3 mg/mL acetone - 35.4 mg/mL ethanol (24 c), 371 mg/mL dmso - 492 mg/mL dmf (24 c)
Formula Weight 479.6
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Cytochalasin B
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Suspected of damaging fertility or the unborn child if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 239-000-2

RUO – Research Use Only