missing translation for 'onlineSavingsMsg'
Learn More

(-)-Cytisine 98.0+%, TCI America™

Catalog No. C2609100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2609100MG 100 mg
C26091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2609100MG Supplier TCI America Supplier No. C2609100MG

Chemical Identifiers

CAS 485-35-8
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD00136048
InChI Key ANJTVLIZGCUXLD-DTWKUNHWSA-N
Synonym cytisine, baptitoxine, sophorine, --cytisine, laburnin, baptitoxin, tabex, cytiton, cytitone, ulexine
PubChem CID 10235
ChEBI CHEBI:4055
SMILES C1C2CNCC1C3=CC=CC(=O)N3C2

Specifications

Melting Point 156°C
Color Yellow
Boiling Point 218°C
Quantity 100 mg
UN Number 2811
Formula Weight 190.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Cytisine
RTECSNumber HA4025000
TSCA Yes