accessibility menu, dialog, popup

UserName

CAS RN 485-35-8

IUPAC Name: (1S,9S)-7,11-diazatricyclo[7.3.1.0²,⁷]trideca-2,4-dien-6-one
Synonyms: cytisine
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: ANJTVLIZGCUXLD-IUCAKERBSA-N
SMILES: O=C1C=CC=C2[C@@H]3CNC[C@H](C3)CN12
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)