missing translation for 'onlineSavingsMsg'
Learn More

Cytisine, 98%, Thermo Scientific™

$209.48 - $740.00

Chemical Identifiers

CAS 485-35-8
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.24
InChI Key ANJTVLIZGCUXLD-DTWKUNHWSA-N
Synonym cytisine, baptitoxine, sophorine, --cytisine, laburnin, baptitoxin, tabex, cytiton, cytitone, ulexine
PubChem CID 10235
ChEBI CHEBI:4055
SMILES C1C2CNCC1C3=CC=CC(=O)N3C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC458852500
View Documents Promotion Details
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC458850010
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

485-35-8
190.24
cytisine, baptitoxine, sophorine, --cytisine, laburnin, baptitoxin, tabex, cytiton, cytitone, ulexine
CHEBI:4055
C11H14N2O
ANJTVLIZGCUXLD-DTWKUNHWSA-N
10235
C1C2CNCC1C3=CC=CC(=O)N3C2

Specifications

485-35-8
5% max.
97.5% min. (HPLC)
cytisine, baptitoxine, sophorine, --cytisine, laburnin, baptitoxin, tabex, cytiton, cytitone, ulexine
ANJTVLIZGCUXLD-DTWKUNHWSA-N
190.24
CHEBI:4055
98%
218°C (3.0 hPa)
Authentic
C11H14N2O
Solubility in water: 493g/L (16°C).
C1C2CNCC1C3=CC=CC(=O)N3C2
10235
190.24
Cytisine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy

GHS Signal Word: Danger

EINECSNumber : 207-616-

SDS
Documents

Documents

RUO – Research Use Only