missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytidine 98.0+%, TCI America™
$32.28 - $163.28
Chemical Identifiers
| CAS | 65-46-3 |
|---|---|
| Molecular Formula | C9H13N3O5 |
| Molecular Weight (g/mol) | 243.22 |
| MDL Number | MFCD00006545 |
| InChI Key | UHDGCWIWMRVCDJ-UHFFFAOYNA-N |
| Synonym | cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone |
| PubChem CID | 6175 |
| ChEBI | CHEBI:17562 |
| IUPAC Name | 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one |
| SMILES | NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O |
Chemical Identifiers
| 65-46-3 | |
| 243.22 | |
| UHDGCWIWMRVCDJ-UHFFFAOYNA-N | |
| 6175 | |
| 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one |
| C9H13N3O5 | |
| MFCD00006545 | |
| cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone | |
| CHEBI:17562 | |
| NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O |
Specifications
| 65-46-3 | |
| C9H13N3O5 | |
| 1 g | |
| UHDGCWIWMRVCDJ-UHFFFAOYNA-N | |
| 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one | |
| 6175 | |
| 243.22 | |
| Crystalline Powder |
| White | |
| MFCD00006545 | |
| cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone | |
| NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O | |
| 243.22 | |
| CHEBI:17562 | |
| ≥98.0% (HPLC,T) | |
| Cytidine |
Safety and Handling
EINECSNumber : (5)-0941
RTECSNumber : UW7370000
TSCA : Yes