missing translation for 'onlineSavingsMsg'
Learn More

Cytidine Sulfate 98.0+%, TCI America™

Catalog No. C05251G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C05251G 1 g
C0525100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C05251G Supplier TCI America Supplier No. C05251G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32747-18-5
Molecular Formula C9H15N3O9S
Molecular Weight (g/mol) 341.291
MDL Number MFCD00151091
InChI Key SYPYJHGPUCBHLU-IAIGYFSYSA-N
Synonym cytidine; sulfuric acid
PubChem CID 21120204
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;sulfuric acid
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O.OS(=O)(=O)O

Specifications

Melting Point 225°C
Color White
Quantity 1 g
Formula Weight 584.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cytidine Sulfate