accessibility menu, dialog, popup

UserName

CAS RN 32747-18-5

IUPAC Name: 4-amino-1-[(2S,3S,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one; sulfuric acid
Synonyms: 4-amino-1-[(2S,3S,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one; sulfuric acid
Molecular Weight (g/mol): 341.29
Molecular Formula: C9H15N3O9S
InChi Key: SYPYJHGPUCBHLU-MTNQCOOWSA-N
SMILES: OS(O)(=O)=O.NC1=NC(=O)N(C=C1)[C@H]1O[C@@H](CO)[C@H](O)[C@@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results