missing translation for 'onlineSavingsMsg'
Learn More

Cytidine, free base, 99.8%, MP Biomedicals™

Catalog No. ICN10145905
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10145905 5 g
ICN10145910 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10145905 Supplier MP Biomedicals Inc Supplier No. 0210145905

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

Specifications

Color White
Assay Percent Range 99.8%
Quantity 5 g
Formula Weight 243.22
Physical Form Crystalline Powder
Chemical Name or Material Cytidine, free base