missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine, 99%

Catalog No. AAA1026109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1026109 10 g
AAA1026118 50 g
AAA1026130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1026109 Supplier Thermo Scientific Chemicals Supplier No. A1026109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Used to synthesize enzyme inhibitors, antiviral agents, and anticancer agents. And also used as constituent of nucleic acids. | CAS: 65-46-3 | C9H13N3O5 | 243.22 g/mol

Cytidine is used to synthesize enzyme inhibitors, antiviral agents, and anticancer agents. And also used as constituent of nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
MDL Number MFCD00006545
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

Specifications

Melting Point ∼215°C (decomposition)
Packaging Plastic bottle
Quantity 10 g
Beilstein 89173
Sensitivity Hygroscopic
Solubility Information Soluble in water and dimethyl sulfoxide.
Formula Weight 243.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Cytidine
EINECSNumber 200-610-9
RTECSNumber UW7370000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only