missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine, 99+%

Catalog No. AC111810100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111810100 10 g
AC111810500 50 g
AC111812500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111810100 Supplier Thermo Scientific Chemicals Supplier No. 111810100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65-46-3 | C9H13N3O5 | 243.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
MDL Number MFCD00006545
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

Specifications

Absorbance 0.43 to 0.47 (A250/A260), 2.05 to 2.15 (A280/A260)
Melting Point 210.0°C to 217.0°C
Color White
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 31,24
Merck Index 15,2783
Specific Rotation + 31.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acids and alkalies,insoluble in organic solvents
Spectrophotometry 98 to 102% (lambda max. 280nm,pH 1) molar absorptivity 13000+/-2%,on dry basis
Transmittance 98% min. (c=1, H2O, 430nm) 1cm cell
Formula Weight 243.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Cytidine, 99.5%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 200-610-9
RTECSNumber UW7370000
TSCA TSCA

RUO – Research Use Only