Learn More
Thermo Scientific Chemicals Cytidine-5'-diphosphocholine sodium salt
Intermediate in the synthesis of phosphatidylcholine; also a phospholipase inhibitor | CAS: 33818-15-4 | C14H25N4NaO11P2 | 510.31 g/mol
Supplier: Thermo Scientific Chemicals J6416114
Description
Cytidine-5'-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCytidine-5′-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.
Solubility
Soluble in water (100 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| Cytidine-5'-diphosphocholine sodium salt hydrate | |
| White | |
| MFCD00150508 | |
| 4110040 | |
| Soluble in water (100mg/ml). | |
| [Na+].C[N+](C)(C)CCOP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O | |
| 510.31 | |
| 510.31 (Anhydrous) |
| 33818-15-4 | |
| C14H25N4NaO11P2 | |
| 25 g | |
| citicolinesodium, cytidine 5'-diphosphocholine sodium, cdp-choline sodium, citicoline, sodium, cdp-choline, sodium, cytidine 5'-diphosphocholine sodium salt, cytidine 5'-diphosphocholine sodium salt dihydrate, cytidine-5'-diphosphocholine monosodium salt, sodium 2s,3r,4s,5s-5-4-amino-2-oxo-1-pyrimidinyl-3,4-dihydroxy-2-oxolanyl methoxy-oxidophosphoryl 2-trimethylammonio ethyl phosphate, sodium 2s,3r,4s,5s-5-4-azanyl-2-oxidanylidene-pyrimidin-1-yl-3,4-bis oxidanyl oxolan-2-yl methoxy-oxidanidyl-phosphoryl 2-trimethylazaniumyl ethyl phosphate | |
| YWAFNFGRBBBSPD-OCMLZEEQSA-M | |
| sodium;[[(2S,3R,4S,5S)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] 2-(trimethylazaniumyl)ethyl phosphate | |
| 46738085 | |
| Powder |
Chemical Identifiers
| 33818-15-4 | |
| 510.31 | |
| YWAFNFGRBBBSPD-OCMLZEEQSA-M | |
| 46738085 | |
| [Na+].C[N+](C)(C)CCOP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O |
| C14H25N4NaO11P2 | |
| MFCD00150508 | |
| citicolinesodium, cytidine 5'-diphosphocholine sodium, cdp-choline sodium, citicoline, sodium, cdp-choline, sodium, cytidine 5'-diphosphocholine sodium salt, cytidine 5'-diphosphocholine sodium salt dihydrate, cytidine-5'-diphosphocholine monosodium salt, sodium 2s,3r,4s,5s-5-4-amino-2-oxo-1-pyrimidinyl-3,4-dihydroxy-2-oxolanyl methoxy-oxidophosphoryl 2-trimethylammonio ethyl phosphate, sodium 2s,3r,4s,5s-5-4-azanyl-2-oxidanylidene-pyrimidin-1-yl-3,4-bis oxidanyl oxolan-2-yl methoxy-oxidanidyl-phosphoryl 2-trimethylazaniumyl ethyl phosphate | |
| sodium;[[(2S,3R,4S,5S)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] 2-(trimethylazaniumyl)ethyl phosphate |
Safety and Handling
EINECSNumber : 251-689-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only